missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Quanti-Cult Plus™ Convenience Set, Nutritional Adequacy Testing QC Set

Product Code. 12936047
Change view
Click to view available options
Quantity:
120 Tests/Set
Unit Size:
120 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12936047 120 Tests/Set 120 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
If you would like to order, a signed ATCC agreement is required.
Product Code. 12936047 Supplier Thermo Scientific™ Supplier No. R4723000

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
If you would like to order, a signed ATCC agreement is required.

Supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.

Thermo Scientific™ Quanti-Cult Plus Nutritional Adequacy Testing Set is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.

  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1mL inoculum

Includes:

12 preserved organism vials (two per organism); 12 rehydration vials

Contents:

Aspergillus brasiliensis ATCC™ 16404™

Bacillus spizizenii ATCC™ 6633 

Candida albicans ATCC™ 10231™

Escherichia coli ATCC™8739™

Pseudomonas paraeruginosa ATCC™ 9027™

Staphylococcus aureus subsp. aureus ATCC™ 6538™.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® 16404™
Description Nutritional Adequacy Testing QC Set/6
Format Quanti-Cult Plus
Product Type QC set
Target Organism Class Organism Sets
Test Type Quantitative QC
Packaging Type 120 Test/Set
Quantity 120 Tests/Set

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.