missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Quanti-Cult Plus™ Convenience Set, Growth Promotion Testing 2 QC Set

Product Code. 12926047
Click to view available options
Quantity:
Set of 2, 100 Tests/Set
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 12926047

Brand: Thermo Scientific™ R4721001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.

Thermo Scientific™ Quanti-Cult Plus Growth Promotion Testing set is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.

  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1mL inoculum

Includes:

10 preserved organism vials (two per organism); 12 rehydration vials

Contents:

  • Aspergillus brasiliensis ATCC™ 16404™†
  • Bacillus subtilis ATCC™ 6633™†
  • Candida albicans ATCC™ 10231™†
  • Pseudomonas aeruginosa ATCC™ 9027™†
  • Staphylococcus aureus subsp. aureus ATCC™ 6538™†
TRUSTED_SUSTAINABILITY

Specifications

ATCC® 16404TM*
Description Growth Promotion Testing 2 QC Set/5
Format Quanti-Cult Plus
Product Type Culti-Loops Set
Target Organism Class Organism Sets
Test Type Quantitative QC
Packaging Type 100 Test/Set
Quantity Set of 2, 100 Tests/Set

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.