missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Quanti-Cult™ Bacillus spizizenii ATCC™ 6633™

Product Code. 12677689
Change view
Click to view available options
Description:
Bacillus spizizenii ATCC™ 6633™
Unit Size:
10 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
12677689 Bacillus spizizenii ATCC™ 6633™ 10 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12677689 Supplier Thermo Scientific™ Supplier No. R4731221
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.

Thermo Scientific™ Quanti-Cult™ Bacillus spizizenii ATCC™ 6633™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial, ready for rehydration and use in quality control procedures.

  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.3mL inoculum

Ordering Alerts:

1 Test/reconstituted vial, 10 Tests/kit

Includes:

10 preserved organism vials and 10 rehydration vials.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® 6633TM*
Description Bacillus spizizenii ATCC™ 6633™
Format Quanti-Cult
Product Type Bactidisk
Target Organism Class Gram-positive Rods
Test Type Quantitative QC
Packaging Type 10 Test/Kit

*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.