Learn More
Thermo Scientific™ Quant™ Total Hardness Test Strips
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Quantity | 1 Kit |
| Range | 50 to 410mg/L (ppm) or 5 to 26°e |
| Storage Requirements | Ambient temperatures |
| Type | Total Hardness Test Strips |