missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Quant-iT™ RNA HS Reagent

Product Code. 15401244
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15401244 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15401244 Supplier Invitrogen™ Supplier No. Q32884
  • ONLINE EXCLUSIVE PRICE
    £634.00
    £595.65 / 1mL
    Save £38.35
    6% Off
View all promotions
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quant-iT™ RNA HS Reagent

The Quant-iT™ RNA HS Reagent makes RNA quantitation easy and accurate, with a linear detection range of 5–100 ng in sample volumes of 1–20μl and selectivity for RNA, even in the presence of an equal mass of DNA. Using the protocol provided, this concentrated assay reagent provides sufficient material to perform at least 2000 assays in a 96-well microplate format. The assay is performed at room temperature, and the signal is stable for approximately 3 hours. Common contaminants, such as salts, solvents, detergents, and protein are well tolerated in the assay.

Important Features of Quant-iT™ RNA HS Reagent:

  • Accurately detects as little as 2 ng RNA in a microplate well
  • Core dynamic range of 5–100 ng
  • Approximate fluorescence excitation/emission maxima of 644/673 nm

    Additional Options for RNA Quantitation:
     
  • For 1–20 samples: use Qubit™ RNA Assay Kit with the Qubit™ Fluorometer
  • For 20–2,000 samples: use Quant-iT™ RNA Assay Kit with fluorescent microplate readers

    Discover the Qubit™ Fluorometer
    The Qubit™ 2.0 Fluorometer is Life Technologies’ second generation fluorometer designed to work seamlessly with our highly specific and sensitive Qubit™ quantitation assays as well as the assays you create with the new MyQubit feature.

    Introducing MyQubit
    New firmware called 'MyQubit' allows you to create applications in minutes using parameters that can easily be uploaded to the instrument using a USB drive, without changing the existing assays. All new Qubit™ 2.0 instruments are pre-loaded with the new MyQubit firmware, and existing owners can download the firmware here: V3.10 firmware download. Any reagent or assay that is spectrally compatible with the Qubit™ hardware can be adapted for use with the Qubit™ 2.0 Fluorometer. Additionally we have developed assays that are ready to download, including glucose and cholesterol assays for research and drug discovery applications.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Quant-iT™ RNA HS Reagent, 1mL, FFPE & Fixed Samples, RNA Sample Type, Store at 2 to 8°C, For Use With Fluorometer, Microplate Reader
Density 0.57 at 645nm
Sample Type Tissue (FFPE), Total RNA, mRNA
Formulation Liquid
For Use With (Application) DNA & RNA purification & analysis, nucleic acid quantitation, RNA quantitation
For Use With (Equipment) Fluorometer, microplate reader
Product Type Quant-iT™ RNA HS Reagent
Quantity 1 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.