Learn More
Invitrogen™ Quant-iT™ RiboGreen ReadyPlate
Description
The Quant-iT RiboGreen ReadyPlate is a preloaded 96-well plate that offers a rapid, simplified, and highly sensitive method for RNA quantification. The ReadyPlate is prefilled with buffer and RiboGreen reagent, which exhibits bright green fluorescence upon binding to RNA or DNA.
The streamlined assay format, along with prediluted standards, enables users to get to their results faster. Here, the user needs to simply hydrate the wells, add the provided standards and user samples, and incubate for two minutes prior to fluorescent measurement. This format allows users to quantify 1–200 ng of RNA (0.01–2 ng/μL when using 100 μL samples).
The Quant-iT RiboGreen ReadyPlate is ideal for measuring yields of RNA preparations for microarray experiments, reverse transcription PCR (RT-PCR), differential display PCR, northern blot analysis, S1 nuclease assays, RNase protection assays, cDNA library preparations, and high-throughput automated workflows.
Specifications
Specifications
| Detection Method | Fluorescence |
| Format | Kit |
| Label or Dye | Other Labels or Dyes |
| Final Product Type | RNA (Labeled) |
| Sample Type | RNA |
| Labeling Target | RNA (General) |
| Product Type | Pre-filled Microplate |
| Quantity | 1 x 96-Well ReadyPlate |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.