missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Quant-iT™ PicoGreen ReadyPlate

Product Code. 30284697
Change view
Click to view available options
Quantity:
1 x 96-Well ReadyPlate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30284697 1 x 96-Well ReadyPlate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30284697 Supplier Invitrogen™ Supplier No. P32705
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Quant-iT PicoGreen ReadyPlate is a preloaded 96-well plate that offers a rapid, ultra-sensitive, and selective method for double-stranded DNA quantification. The plate is prefilled with buffer and PicoGreen reagent, which exhibits bright green fluorescence upon binding to DNA.

The streamlined assay protocol requires users to simply hydrate the wells, add the provided standards and user samples, and incubate for two minutes prior to fluorescent measurement. This format allows users to quantify 0.2–200 ng of DNA (0.002–2 ng/μL when using 100 μL samples).

The Quant-iT PicoGreen ReadyPlate is ideal for quantitating small amounts of DNA for many applications, including synthesizing cDNA for library production, quantifying DNA libraries for next-generation sequencing, and purifying DNA fragments for subcloning.

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Fluorescence
Format Kit
Label or Dye Other Labels or Dyes
Final Product Type DNA
Sample Type DNA
Labeling Target DNA
Product Type Pre-filled Microplate
Quantity 1 x 96-Well ReadyPlate

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.