missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Quant-iT™ microRNA Reagent

Product Code. 15491234
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 15491234

Brand: Invitrogen™ Q32883

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Quant-iT™ microRNA Reagent

The Quant-iT™ microRNA Reagent allows easy and accurate quantification of microRNA (miRNA), even in the presence of common contaminants such as salts, free nucleotides, solvents, detergents, and protein. The assay is highly selective for miRNA over rRNA or large mRNA (>1000 bp). Although the reagent is not exclusively selective for miRNA, we have been able to reproducibly quantify miRNA in pure samples at levels as low as 0.5 ng following the supplied protocol.

Features of Quant-iT™ microRNA Reagent:

  • Accurately detects as little as 0.5 ng miRNA in a microplate well
  • Core dynamic range of 5–500 ng/mL
  • Accurate results for sample concentrations from 50 ng/mL to 100μg/mL
  • Approximate fluorescence excitation/emission maxima of 500/525 nm

    Using the protocol provided, this concentrated assay reagent provides sufficient material to perform at least 2000 assays in a 96-well microplate format. The assay is performed at room temperature, and the signal is stable for approximately 3 hours. Accurate results can be obtained for initial sample concentrations ranging from 50 ng/mL–100μg/mL.

    Which product to choose for microRNA Quantitation?

  • For 1–20 samples: use the Qubit™ microRNA Assay Kit with the Qubit™ Fluorometer
  • For 20–2,000 samples: use the Quant-iT™ microRNA Assay Kit with microplate reader
  • For >2,000 samples: use this Quant-iT™ microRNA Reagent with microplate reader

    Discover the Qubit™ Fluorometer
    The Qubit™ 2.0 Fluorometer is Life Technologies’ second generation fluorometer designed to work seamlessly with our highly specific and sensitive Qubit™ quantitation assays as well as the assays you create with the new MyQubit feature.

    Introducing MyQubit
    New firmware called 'MyQubit' allows you to create applications in minutes using parameters that can easily be uploaded to the instrument using a USB drive, without changing the existing assays. All new Qubit™ 2.0 instruments are pre-loaded with the new MyQubit firmware, and existing owners can download the firmware here: V3.10 firmware download. Any reagent or assay that is spectrally compatible with the Qubit™ hardware can be adapted for use with the Qubit™ 2.0 Fluorometer. Additionally we have developed assays that are ready to download, including glucose and cholesterol assays for research and drug discovery applications.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage 2°C to 8°C
    Description Quant-iT™ microRNA Reagent, 1mL, FFPE & Fixed Samples, RNA Sample Type, Store at 2 to 8°C, For Use With Fluorometer, Microplate Reader
    Density 0.67 at 494nm
    Sample Type Tissue (FFPE), Total RNA, mRNA
    Formulation Liquid
    For Use With (Application) DNA & RNA purification & analysis, nucleic acid quantitation, RNA quantitation
    For Use With (Equipment) Fluorometer, microplate reader
    Product Type Quant-iT™ microRNA Reagent
    Quantity 1 mL

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.