Learn More
Sartorius Stedim Biotech™ Qualitative/Technical Papers, Smooth, Folded, Grade 3 m/N
Sartorius Stedim Biotech™ Qualitative/Technical Papers, Smooth, Folded, Grade 3 m/N are suitable for analytical and technical purposes where medium fast filtering is desirable.
Brand: Sartorius Stedim Biotech™ FT-4-305-230
Additional Details : Weight : 0.35800kg
Description
- Made of refined pulp and linters
- Medium fast filtering
- Non-sterile and bulk packed
Specifications
Folded | |
Cotton Linters and Refined Pulp (>95% alpha-cellulose) | |
0.14 mm | |
65 g/m2 |
7 to 10μm | |
230 mm | |
3 m/N | |
100/Pk. |