missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Qualitative Analysis Test Papers, MN 615

Product Code. 10176087 Shop All Macherey-Nagel Products
Change view
Click to view available options
Diameter (Metric):
10 mm
15 mm
20 mm
25 mm
30 mm
82 mm
85 mm
Unit Size:
Pack of 100
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10176087 85 mm Pack of 100
10758460 10 mm Pack of 100
10391255 15 mm Pack of 100
10187374 20 mm Pack of 100
10499634 25 mm Pack of 100
10320415 30 mm Pack of 100
10402275 82 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10176087 Supplier Macherey-Nagel™ Supplier No. 4310085
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Widely used for routine separations.

  • Qualitative filter papers widely used for routine separations
  • Manufactured from top quality raw materials using ultra pure water to ensure consistent and reliable filtration
  • Most widely used general purpose filter paper with medium retention and flow rate
  • This filter paper covers a wide range of laboratory applications and is frequently used for clarifying liquids

Specifications

Format Disc
Grade MN 615
Quantity 100/Pk.
Diameter (Metric) 85 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.