missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Qualitative filter papers widely used for routine separations
- Manufactured from top quality raw materials using ultra pure water to ensure consistent and reliable filtration
- Most widely used general purpose filter paper with medium retention and flow rate
- This filter paper covers a wide range of laboratory applications and is frequently used for clarifying liquids
Specifications
Specifications
| Format | Disc |
| Grade | MN 615 |
| Quantity | 100/Pk. |
| Diameter (Metric) | 10 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?