Learn More
BINDER™ Qualification Documents IQ/OQ
Product Code. 16494009 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers (Series BD, BF, ED, FD, FED, FDL, FP, M, C, CB, KB, KT, MK, MKT, VD, VDL, UF V)
BINDER Chambers (Series KBF, KBF-S, KMF, MKF, MKFT)
Unit Size:
Each
Product Code. 16494009
Brand: BINDER™ 70570002
Qualification documents - Digital in PDF format, IQ/OQ documents – supporting documents for validation performed by customers, consisting of: IQ/OQ checklists including calibration guide and comprehensive unit documentation.
  • Parameters depending on unit
  • Available as hardcopy inside folder or PDF document
Specifications
For Use With (Equipment) BINDER Chambers (Series KBF, KBF-S, KMF, MKF, MKFT)
Description Qualification documents - Digital in PDF format, IQ/OQ documents - supporting documents for validation performed by customers, consisting of: IQ/OQ checklists incl. calibration guide and comprehensive unit documentation; parameters: temperature and humidity values
Product Type IQ/OQ Documentation (PDF)
Product Line BINDER Chambers
Service Offering IQ/OQ/PQ with completion of documents