missing translation for 'onlineSavingsMsg'
Learn More

Quadrol, MedChemExpress

Product Code. 16468738
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468738 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16468738 Supplier Medchem Express Supplier No. HYB2149
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Quadrol is an immunostimulant and has been implicated as a potentially useful agent in accelerated wound healing.

Quadrol is an immunostimulant and has been implicated as a potentially useful agent in accelerated wound healing.

Specifications

Chemical Name or Material Quadrol
CAS 102-60-3
Color Colorless
Purity Grade Notes Research
Molecular Formula C14H32N2O4
Quantity 10 g
Synonym N,N,N′,N′-Tetrakis(2-hydroxypropyl)ethylenediamine EDTP
Solubility Information Ethanol : 100 mg/mL (341.99 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (341.99 mM; Need ultrasonic)
SMILES CC(O)CN(CC(O)C)CCN(CC(O)C)CC(O)C
Molecular Weight (g/mol) 292.41
Formula Weight 292.41
Percent Purity 98.0%
Grade Research
Physical Form Oil
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.