Learn More
Honeywell™ QUAD (i)XTREM S3 Shoes
Product Code. 15856595
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15856595 37 1 pair
15866595 38 1 pair
15876595 39 1 pair
15886595 40 1 pair
15896595 41 1 pair
15806605 42 1 pair
15816605 43 1 pair
15826605 44 1 pair
15836605 45 1 pair
15846605 46 1 pair
15856605 47 1 pair
15866605 48 1 pair
15876605 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15856595 Supplier Honeywell™ Supplier No. 624621237/7
Only null left
Add to basket
Add to basket
Honeywell™ QUAD (i)XTREM S3 Shoes are constructed of water-resistant full grain leather.
  • Water resistant grain full leather
  • Comfort foam for improved foot stability
  • Gusset to avoid water and dust penetration
  • Poromax 3D lining
  • Toe cap metal free
  • Metal free antiperforation insert
  • Injected PU / Nitrile sole
  • (i)NTELLIGENT wear control
  • EN ISO 20345:2011 S3 HRO HI CI SRC
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN ISO 20345:2011 S3 HRO HI CI SRC
Color Brown, Black
Material Leather
Size 37
Sole Material Polyurethane and nitrile
Toe Style Metal-free