missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ QSOX2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA534150

Product Code. 15352791

  • £442.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Percent identity with other species by BLAST analysis: Human (100%) Gorilla (95%).

QSOX2 is a member of the sulfhydryl oxidase/quiescin-6 (Q6) family (QSOX1) that regulates the sensitization of neuroblastoma cells for IFN-gamma (IFNG)-induced cell death.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

QSOX2
Polyclonal
Unconjugated
Qsox2
BC030934; neuroblastoma-derived sulfhydryl oxidase; QSCN6L1; QSOX2; quiescin Q6 sulfhydryl oxidase 2; quiescin Q6-like 1; quiescin Q6-like protein 1; quiescin sulfhydryl oxidase 2; SOXN; Sulfhydryl oxidase 2; thiol oxidase 2
Rabbit
Antigen Affinity Chromatography
RUO
169714
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.5 mg/mL
PBS with 0.1% sodium azide
Q6ZRP7
Qsox2
Synthetic 19 amino acid peptide from internal region of human QSOX2.
50 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.