missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QSOX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12713-1-AP-150UL
This item is not returnable.
View return policy
Description
The QSOX1 gene, also known as Quiescin Q6, is a fusion of two ancient genes: thioredoxin and ERV1. Its expression is induced as fibroblasts begin to exit the proliferative cycle and enter quiescence, suggesting that this gene plays an important role in growth regulation. The QSOX1 protein oxidizes sulfhydryl groups to form disulfide bonds in proteins. QSOX1 expression is induced by hypoxia and appears to protect cells against oxidative stress-induced apoptosis.Specifications
| QSOX1 | |
| Polyclonal | |
| Unconjugated | |
| QSOX1 | |
| hQSOX, Q6, QSCN6, QSOX1, Quiescin Q6, Sulfhydryl oxidase 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 104009, 5768, 84491 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O00391, Q6IUU3, Q8BND5 | |
| Qsox1 | |
| QSOX1 Fusion Protein Ag3396 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction