missing translation for 'onlineSavingsMsg'
Learn More

QSOX1/Quiescin Q6 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18413165 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413165 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413165 Supplier Novus Biologicals Supplier No. NBP281797AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

QSOX1/Quiescin Q6 Polyclonal antibody specifically detects QSOX1/Quiescin Q6 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen QSOX1/Quiescin Q6
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias EC 1.8.3.2, FLJ34858, hQSOX, Q6, QSCN6, Quiescin Q6, quiescin Q6 sulfhydryl oxidase 1, sulfhydryl oxidase 1
Host Species Rabbit
Immunogen QSOX1 antibody was raised against a 17 amino acid peptide near the center of human QSOX1. The immunogen is located within amino acids 360 - 410 of QSOX1. Amino Acid Squence: FKTALDDRKEGAVLAKK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5768
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.