missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ QSOX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566006
This item is not returnable.
View return policy
Description
Immunogen sequence: CSACHNERLDV PVWDVEATLN FLKAHFSPSN IILDFPAAGS AARRDVQNVA AAPELAMGAL ELESRNSTLD P Highest antigen sequence identity to the following orthologs - mouse 58%, rat 59%.
The QSOX1 gene, also known as Quiescin Q6, is a fusion of two ancient genes: thioredoxin and ERV1. Its expression is induced as fibroblasts begin to exit the proliferative cycle and enter quiescence, suggesting that this gene plays an important role in growth regulation. The QSOX1 protein oxidizes sulfhydryl groups to form disulfide bonds in proteins. QSOX1 expression is induced by hypoxia and appears to protect cells against oxidative stress-induced apoptosis.
Specifications
| QSOX1 | |
| Polyclonal | |
| Unconjugated | |
| Qsox1 | |
| 1300003H02Rik; FAD-dependent sulfhydryl oxidase-2; hQSOX; mSOx; Q6; Qscn6; Qsox; QSOX1; Quiescin Q6; quiescin Q6 sulfhydryl oxidase 1; quiescin sulfhydryl oxidase 1; RP11-502H18.3; rQSOX; rSOx; Skin sulfhydryl oxidase; Sox; Sox2; Sox-2; Sulfhydryl oxidase 1; testis tissue sperm-binding protein Li 62n; thiol oxidase 1; UNQ2520/PRO6013 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5768 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00391 | |
| Qsox1 | |
| Recombinant Human QSOX1. Recombinant protein control fragment (Product #RP-105036). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction