missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QS11, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
QS11 is an inhibitor of ARFGAP1 (ADP-ribosylation factor GTPase-activating protein 1), with an EC50 of 1.5 μM. QS11 modulates Wnt/β-catenin signaling through an effect on protein trafficking. QS11 inhibits migration of ARFGAP overexpressing breast cancer cells.
Specifications
Specifications
| Chemical Name or Material | QS11 |
| CAS | 944328-88-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C36H33N5O2 |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 100 mg/mL (176.16 mM) |
| SMILES | OC[C@H](CC1=CC=CC=C1)NC2=C3C(N(CC4=CC=C(C5=CC=CC=C5)C=C4)C=N3)=NC(OC6=CC=C(CCC7)C7=C6)=N2 |
| Molecular Weight (g/mol) | 567.68 |
| Formula Weight | 567.68 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction