missing translation for 'onlineSavingsMsg'
Learn More

QS11, MedChemExpress

Product Code. 16418047
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16418047

Brand: Medchem Express HY12762

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress QS11 is an inhibitor of ARFGAP1 (ADP-ribosylation factor GTPase-activating protein 1), with an EC50 of 1.5 μM. QS11 modulates Wnt/β-catenin signaling through an effect on protein trafficking. QS11 inhibits migration of ARFGAP overexpressing breast cancer cells.

QS11 is an inhibitor of ARFGAP1 (ADP-ribosylation factor GTPase-activating protein 1), with an EC50 of 1.5 μM. QS11 modulates Wnt/β-catenin signaling through an effect on protein trafficking. QS11 inhibits migration of ARFGAP overexpressing breast cancer cells.

Specifications

Chemical Name or Material QS11
CAS 944328-88-5
Color White
Purity Grade Notes Research
Molecular Formula C36H33N5O2
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (176.16 mM)
SMILES OC[C@H](CC1=CC=CC=C1)NC2=C3C(N(CC4=CC=C(C5=CC=CC=C5)C=C4)C=N3)=NC(OC6=CC=C(CCC7)C7=C6)=N2
Molecular Weight (g/mol) 567.68
Formula Weight 567.68
Percent Purity 99.42%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.