missing translation for 'onlineSavingsMsg'
Learn More

Hellma™ Suprasil™ Quartz Ultra-Micro Cell Cuvettes

Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Suprasil™ Quartz Ultra-Micro Cell Cuvettes come with either a PE stopper or open with pipette tips.

Brand:  Hellma™ 105-200-15-40

Code : 33

Additional Details : Weight : 0.02068kg

 View more versions of this product

Product Code. 10446719

  • £320.00 / Each


In Stock 2
Add to basket

Description

Description

  • Very chemically resistant
  • Maintains shape, even at high temperatures up to approximately 1000°C
Specifications

Specifications

160μL
45mm
12.5mm
Ultra Macro Absorption Cells
Open with pipette tips
10mm
QS Quartz Suprasil
Videos
Documentation Special Offers

Special Offers

  • Use code 13096 to receive this promotion.
    : 13096

    2018-05-15

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2018 and 15/05/2018 subject to availability of the product in stock. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy or if the product is no longer available in stock. Other restrictions may apply.

    Terms & Conditions