missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grant Instruments™ QR Racks for Grant Heated Circulating Baths
Description
- Accommodates tubes as small as 0.5mL or as large as 30mm dia.
- For use with Grant Heated Circulating Baths and Heating Circulators.
Specifications
Specifications
| Capacity (English) | 10 tubes |
| For Use With (Equipment) | Grant Heated Circulating Baths |
| Holds | 24 mm Tubes |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?