missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QPRT Antibody (24C4), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59433AF594
This item is not returnable.
View return policy
Description
QPRT Monoclonal antibody specifically detects QPRT in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| QPRT | |
| Monoclonal | |
| Alexa Fluor 594 | |
| EC 2.4.2.19, nicotinate-nucleotide pyrophosphorylase (carboxylating), QAPRTase, QPRTasenicotinate-nucleotide pyrophosphorylase [carboxylating], quinolinate phosphoribosyltransferase, Quinolinate phosphoribosyltransferase [decarboxylating] | |
| Recombinant human QPRT (1-297aa) purified from E. coli | |
| 0.1 mL | |
| Neuroscience | |
| 23475 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA, Immunofluorescence | |
| 24C4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction