missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ qPCR & Go SYBR Lo-ROX Kit
Shop All MP Biomedicals Products
Click to view available options
No. of Reactions:
2000 Reactions
500 Reactions
Unit Size:
2000 reactions
500 reactions
Description
- The qPCR & Go SYBR™Lo-ROX Kit has been developed for fast, highly reproducible real-time PCR and has been validated on commonly used real-time PCR instruments.
- A combination of the latest advances in buffer chemistry and enhancers, together with an antibody-mediated hot-start DNA polymerase system, ensures that the qPCR & Go SYBR™Lo-ROX Kit delivers fast, highly-specific and ultra-sensitive real-time PCR.
- For ease-of-use and added convenience, qPCR & Go SYBR™Lo-ROX is provided as a 2x mastermix containing all the components necessary for real-time PCR, including the SYBR™ Green I dye, dNTPs, stabilisers and enhancers.
- The kit consists of a ready-to-use premix, only primers and template need to be added.
Specifications
Specifications
| Content And Storage | -20°C |
| For Use With (Equipment) | Fast and sensitive qPCR SYBR Master Mix. For most Agilent platforms and some newer ABI platforms. |
| Product Type | qPCR Kit |
| Quantity | 5 x 1 mL |
| For Use With (Application) | Real Time PCR (qPCR) |
| No. of Reactions | 500 Reactions |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction