missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ qPCR & Go Probe No-ROX Kit

Product Code. 15858551 Shop All MP Biomedicals Products
Click to view available options
No. of Reactions:
2000 Reactions
500 Reactions
Unit Size:
2000 reactions
500 reactions
This item is not returnable. View return policy

Product Code. 15858551

Brand: MP Biomedicals™ 11EBI06200

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Fast and sensitive qPCR Probe Master Mix. For all other platforms, e.g. Roche, Bio-Rad, Qiagen, etc.

  • The qPCR & Go Probe No-ROX Kit has been developed for fast, highly reproducible real-time PCR and has been validated on commonly used real-time PCR instruments.
  • The kit has been formulated for use with probe-detection technology, including TaqMan™, Scorpions™ and molecular beacon probes.
  • A combination of the latest advances in buffer chemistry and PCR enhancers, together with a hot-start DNA polymerase, ensures that the qPCR & Go Probe Kit delivers fast, highly-specific and ultra-sensitive real-time PCR.
  • qPCR & Go Probe is provided as a 2x mastermix containing all the components necessary for real-time PCR, including dNTPs, stabilizers and enhancers.
TRUSTED_SUSTAINABILITY

Specifications

Product Type qPCR Kit
Quantity 4 x 5 mL
For Use With (Application) Real Time PCR (qPCR)
No. of Reactions 2000 Reactions

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.