missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ qPCR & Go Probe No-ROX Kit
Shop All MP Biomedicals Products
Click to view available options
No. of Reactions:
2000 Reactions
500 Reactions
Unit Size:
2000 reactions
500 reactions
Description
- The qPCR & Go Probe No-ROX Kit has been developed for fast, highly reproducible real-time PCR and has been validated on commonly used real-time PCR instruments.
- The kit has been formulated for use with probe-detection technology, including TaqMan™, Scorpions™ and molecular beacon probes.
- A combination of the latest advances in buffer chemistry and PCR enhancers, together with a hot-start DNA polymerase, ensures that the qPCR & Go Probe Kit delivers fast, highly-specific and ultra-sensitive real-time PCR.
- qPCR & Go Probe is provided as a 2x mastermix containing all the components necessary for real-time PCR, including dNTPs, stabilizers and enhancers.
Specifications
Specifications
| Concentration | 2X |
| Content And Storage | dNTPs, stabilizers and enhancers. Store at -20°C |
| For Use With (Equipment) | Fast and sensitive qPCR Probe Master Mix. For all other platforms, e.g. Roche, Bio-Rad, Qiagen, etc. |
| Product Type | qPCR Kit |
| Quantity | 5 x 1 mL |
| For Use With (Application) | Real Time PCR (qPCR) |
| No. of Reactions | 500 Reactions |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction