missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ QM-FN-SO3

Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Description
QM-FN-SO3 is a near-infrared aggregation-induced emission active probe for in vivo imaging of amyloid β (Aβ) plaques. Brain penetrant. Excitation and emission maxima (λ) are 488 and 680 nm, respectively.
Specifications
Specifications
| Chemical Name or Material | Sodium 3-(4-(Dicyanomethylene)-2-(2-(5-(4-(dimethylamino)phenyl)thiophen-2-yl)vinyl)quinolin-1(4H)-yl)propane-1-sulfonate |
| CAS | 2316820-94-5 |
| Quantity | 2 mg |
| Target | Fluorescent Probes |
| Molecular Formula | C29H25N4O3S2.Na |
| Purity | 95% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction