missing translation for 'onlineSavingsMsg'
Learn More

QKI/Quaking Antibody (PCRP-QKI-2F10), Biotin, Novus Biologicals™

Product Code. 18383764 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18383764 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18383764 Supplier Novus Biologicals Supplier No. NBP314072B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

QKI/Quaking Monoclonal antibody specifically detects QKI/Quaking in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen QKI/Quaking
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-QKI-2F10
Conjugate Biotin
Formulation PBS
Gene Alias DKFZp586I0923, HKQ, homolog of mouse quaking QKI (KH domain RNA binding protein), Hqk, hqkI, protein quaking, QK, QK1, QK3, quaking homolog, KH domain RNA binding (mouse), RNA binding protein HQK
Host Species Mouse
Immunogen Recombinant full-length human QKI/Quaking protein (Uniprot: Q96PU8)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9444
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.