missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QKI/Quaking Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-41401
This item is not returnable.
View return policy
Description
QKI/Quaking Polyclonal specifically detects QKI/Quaking in Mouse samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| QKI/Quaking | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| DKFZp586I0923, HKQ, homolog of mouse quaking QKI (KH domain RNA binding protein), Hqk, hqkI, protein quaking, QK, QK1, QK3, quaking homolog, KH domain RNA binding (mouse), RNA binding protein HQK | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize all reported isoforms (NP_006766.1; NP_996735.1; NP_996736.1; NP_996737.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:64000 | |
| NP_006766.1 | |
| QKI | |
| Peptide with sequence ETKEKPKPTPDY-C corresponding to N-Terminus according to NP_006766.1, NP_996735.1, NP_996736.1, NP_996737.1. | |
| 0.1 mg | |
| Neuroscience | |
| 9444 | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction