missing translation for 'onlineSavingsMsg'
Learn More

QIAGEN QIAcard FTA Indicating Micro

Product Code. 11999075
Click to view available options
Quantity:
25 Pk.
Unit Size:
Pack of 25
This item is not returnable. View return policy

Product Code. 11999075

Brand: QIAGEN WB120311

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. Indicating QIAcard FTA Micro with 1 sample area per card.

  • QIAcard Indicating FTA Cards include a pink dye that turns white when a colorless sample, e.g. buccal cells or saliva is applied.
  • Stabilization and protection of nucleic acids at room temperature with for spot volumes of 125 μL.
  • Cell lysis on contact, denatures proteins and protects DNA from degradation.

Specifications

Description Collect, stabilize, process, transport, and archive buccal DNA samples with Indicating FTA Micro Cards. With FTA, DNA stays in the matrix.
Sample Type Buccal Cells, Saliva
For Use With (Application) Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology
No. of Samples 1
Product Type Indicating FTA Micro Card
Quantity 25 Pk.
Sample Volume 125 μL
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.