missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ QDPR Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA563632

Product Code. 15771402

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SLAGKNSGMP PGAAAIAVLP VTLDTPMNRK SMPEADFSSW TPLEFLVETF HDWITGKNRP SSGSLIQVVT TEGRTEL Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.

This gene encodes the enzyme dihydropteridine reductase, which catalyzes the NADH-mediated reduction of quinonoid dihydrobiopterin. This enzyme is an essential component of the pterin-dependent aromatic amino acid hydroxylating systems. Mutations in this gene resulting in QDPR deficiency include aberrant splicing, amino acid substitutions, insertions, or premature terminations. Dihydropteridine reductase deficiency presents as atypical phenylketonuria due to insufficient production of biopterin, a cofactor for phenylalanine hydroxylase.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

QDPR
Polyclonal
Unconjugated
QDPR
2610008L04Rik; 6,7-dihydropteridine reductase; D5Ertd371e; Dhpr; dihydropteridine reductase; HDHPR; HPR; PKU2; QDPR; Quinoid dihydropteridine reductase; SDR33C1; Short chain dehydrogenase/reductase family 33C member 1; short chain dehydrogenase/reductase family 33C, member 1; testis secretory sperm-binding protein Li 236P
Rabbit
Antigen affinity chromatography
RUO
5860
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P09417
QDPR
Recombinant protein corresponding to Human QDPR. Recombinant protein control fragment (Product #RP-101362).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.