missing translation for 'onlineSavingsMsg'
Learn More

QDPR Antibody (2D4-1D3), Novus Biologicals™

Product Code. 18329078 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329078 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18329078 Supplier Novus Biologicals Supplier No. H00005860M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

QDPR Monoclonal antibody specifically detects QDPR in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen QDPR
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2D4-1D3
Conjugate Unconjugated
Dilution ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH00576
Gene Alias dhpr, DHPR member 1, quinoid dihydropteridine reductaseFLJ42391
Host Species Mouse
Immunogen QDPR (AAH00576, 1 a.a. ~ 244 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAAAAAAGEARRVLVYGGRGALGSRCVQAFRARNWWVASVDVVENEEASASIIVKMTDSFTEQADQVTAEVGKLLGEEKVDAILCVAGGWAGGNAKSKSLFKNCDLMWKQSIWTSTISSHLATKHLKEGGLLTLAGAKAALDGTPGMIGYGMAKGAVHQLCQSLAGKNSGMPPGAAAIAVLPVTLDTPMNRKSMPEADFSSWTPLEFLVETFHDWITGKNRPSSGSLIQVVTTEGRTELTPAYF
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5860
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.