missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Qdot™ 655 ITK™ Streptavidin Conjugate Kit
Qdot™ 655 ITK™ Streptavidin Conjugate Kit
Brand: Molecular Probes™ Q10021MP
Code : 50
Additional Details : Weight : 0.50000kg
Description
The Qdot™ 655 ITK™ Streptavidin Conjugate Kit provides the Qdot™ streptavidin conjugate as 250μL of a 2μM solution. Unlike the Qdot™ streptavidin conjugates, the Qdot™ ITK™ streptavidin conjugates have the streptavidin covalently attached to the inner amphiphilic coating without a PEG linker. These Qdo™ conjugates can be used for binding biotinylated probes. The kit also includes 30 mL of Qdot™ incubation buffer.Specifications
2 μM | |
Store in refrigerator (2–8°C). | |
ITK™, Qdot™ |
Qdot 655 | |
Room Temperature | |
Streptavidin Conjugate (fluorescent) |
For Research Use Only. Not for use in diagnostic procedures.