missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Qdot™ 565 ITK™ Streptavidin Conjugate Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
The Qdot™ 565 ITK™ Streptavidin Conjugate Kit provides the Qdot™ streptavidin conjugate as 250μL of a 2μM solution. Unlike the Qdot™ streptavidin conjugates, the Qdot™ ITK™ streptavidin conjugates have the streptavidin covalently attached to the inner amphiphilic coating without a PEG linker. These Qdo™ conjugates can be used for binding biotinylated probes. The kit also includes 30 mL of Qdot™ incubation buffer.
Specifications
Specifications
| Conjugate | Qdot 565 |
| Quantity | 1 Kit |
| Content And Storage | Store in refrigerator (2–8°C). |
| Shipping Condition | Room Temperature |
| Product Line | ITK, Qdot |
| Product Type | Conjugate Kit |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction