missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QBRICK/FREM1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
£462.00
Specifications
| Antigen | QBRICK/FREM1 |
|---|---|
| Applications | Western Blot, ELISA, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Form | Purified |
Description
QBRICK/FREM1 Polyclonal antibody specifically detects QBRICK/FREM1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| QBRICK/FREM1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| BNAR, C9orf143chromosome 9 open reading frame 154, C9orf145C9orf154extracellular matrix protein QBRICK, DKFZp686M16108, FLJ25461, FRAS1 related extracellular matrix 1, FRAS1-related extracellular matrix protein 1, Protein QBRICK, TILRR | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human FREM1. The immunogen is located within the last 50 amino acids of FREM1. Amino Acid Squence: TNGRRGPSPRSKLGK | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunocytochemistry | |
| mFluor Violet 610 SE | |
| Rabbit | |
| Cell Biology | |
| 50 mM Sodium Borate | |
| 158326 | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title