missing translation for 'onlineSavingsMsg'
Learn More

QBRICK/FREM1 Antibody, FITC, Novus Biologicals™

Product Code. 18420411 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420411 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420411 Supplier Novus Biologicals Supplier No. NBP177180F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

QBRICK/FREM1 Polyclonal antibody specifically detects QBRICK/FREM1 in Human, Mouse samples. It is validated for ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen QBRICK/FREM1
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias BNAR, C9orf143chromosome 9 open reading frame 154, C9orf145C9orf154extracellular matrix protein QBRICK, DKFZp686M16108, FLJ25461, FRAS1 related extracellular matrix 1, FRAS1-related extracellular matrix protein 1, Protein QBRICK, TILRR
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human FREM1. The immunogen is located within the last 50 amino acids of FREM1. Amino Acid Squence: TNGRRGPSPRSKLGK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 158326
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.