missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
QBRICK/FREM1 Polyclonal antibody specifically detects QBRICK/FREM1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | QBRICK/FREM1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | BNAR, C9orf143chromosome 9 open reading frame 154, C9orf145C9orf154extracellular matrix protein QBRICK, DKFZp686M16108, FLJ25461, FRAS1 related extracellular matrix 1, FRAS1-related extracellular matrix protein 1, Protein QBRICK, TILRR |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1880-2179 of human FREM1 (NP_659403.4). GSSSSTTSGSFHLERRPLPSSMQLAVIRGDTLRGFDSTDLSQRKLRTRGNGKTVRPSSVYRNGTDIIYNYHGIVSLKLEDDSFPTHKRKAKVSIISQPQKTIKVAELPQADKVESTTDSHFPRQDQLPSFPKNCTLELKGLFHFEEGIQKLYQCNGIAWKAWSPQTKDVEDKSCPAGWHQHSGYCHILITEQKGTWNAAAQACREQYLGNLVTVFSRQHMRWLWDIGGRKSFWIGLNDQVHAGHWEWIGGEPVAFTNGRRGPSQRSKLGKSCVLVQRQGKWQTKDCRRAKPHNYVCSRKL |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?