missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QBRICK/FREM1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94701-0.1ml
This item is not returnable.
View return policy
Description
QBRICK/FREM1 Polyclonal antibody specifically detects QBRICK/FREM1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| QBRICK/FREM1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| BNAR, C9orf143chromosome 9 open reading frame 154, C9orf145C9orf154extracellular matrix protein QBRICK, DKFZp686M16108, FLJ25461, FRAS1 related extracellular matrix 1, FRAS1-related extracellular matrix protein 1, Protein QBRICK, TILRR | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1880-2179 of human FREM1 (NP_659403.4). GSSSSTTSGSFHLERRPLPSSMQLAVIRGDTLRGFDSTDLSQRKLRTRGNGKTVRPSSVYRNGTDIIYNYHGIVSLKLEDDSFPTHKRKAKVSIISQPQKTIKVAELPQADKVESTTDSHFPRQDQLPSFPKNCTLELKGLFHFEEGIQKLYQCNGIAWKAWSPQTKDVEDKSCPAGWHQHSGYCHILITEQKGTWNAAAQACREQYLGNLVTVFSRQHMRWLWDIGGRKSFWIGLNDQVHAGHWEWIGGEPVAFTNGRRGPSQRSKLGKSCVLVQRQGKWQTKDCRRAKPHNYVCSRKL | |
| 0.1 mL | |
| Cell Biology | |
| 158326 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction