missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ QB Series Safety Cover

Product Code. 11450968
Change view
Click to view available options
For Use With (Equipment):
4 Block Systems
For 1 Block Systems
For 2 Block Systems
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11450968 For 2 Block Systems Each
11440968 For 1 Block Systems Each
10097362 4 Block Systems Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450968 Supplier Grant Instruments™ Supplier No. QBL2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Made from tough clear acrylic for maximum visibility. Grant Instruments™ QB Series Safety Covers prevent accidental touching of a hot block or contamination of samples from splashes.

  • For use with QB Series Dry Block Heaters
  • Acrylic construction provides increased visibility
  • Prevents burning from accidentally touching the hot block
  • Clearance height of 85mm
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
For Use With (Equipment) For 2 Block Systems
Material Acrylic
Type Safety Cover
For Use With (Application) For maximum visibility whilst preventing accidental touching of a hot block or contamination of samples from splashes
Height (Metric) 85 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.