missing translation for 'onlineSavingsMsg'
Learn More

QA1b Antibody (6A8.6F10.1A6), HRP, Novus Biologicals™

Product Code. 18400552 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400552 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400552 Supplier Novus Biologicals Supplier No. NBP226649H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

QA1b Monoclonal antibody specifically detects QA1b in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen QA1b
Applications Western Blot, Flow Cytometry, Immunoprecipitation, Neutralization
Classification Monoclonal
Clone 6A8.6F10.1A6
Conjugate HRP
Formulation PBS
Gene Alias H2-Q1, H2-T23
Host Species Mouse
Immunogen Animals were immunized with a synthetic peptide from the alpha-2 domain of Qa-1b. Qa-1 is a Class Ib molecule recognized by cytotoxic T cells and natural killer cells. Spleen cells were fused with P3X63Ag8.653 myeloma cells.The antibody reacts with the Qa-1b Class Ib molecule.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 15040
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.