missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Qa-1(b) Mouse anti-Mouse , PE, Clone: 6A8.6F10.1A6, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566640
This item is not returnable.
View return policy
Description
Qa-1(b) Monoclonal antibody specifically detects Qa-1(b) in Mouse samples. It is validated for Flow Cytometry
Specifications
| Qa-1(b) | |
| Monoclonal | |
| PE | |
| H2-Qa1b; H2-T23; Qa-1(b); Qa-1b; Qa1b; T23b | |
| Qa-1[b] aa. 161-179 Peptide | |
| RUO | |
| Mouse | |
| IgG1 κ |
| Flow Cytometry | |
| 6A8.6F10.1A6 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 25 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction