missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Q-VD-OPH, MP Biomedicals™

Product Code. p-7083071 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 mg
3 mg
5 mg
Unit Size:
1mg
3mg
5mg
This item is not returnable. View return policy

Product Code. 11465386

Brand: MP Biomedicals™ 03OPH10901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifically designed for in-vivo research. Q-VD-OPH, MP Biomedicals is synthesized using proprietary technology as a cell-permeable, irreversible caspase inhibitor. The OPH trap, with its superior potency, proven cell permeability and minimal toxicity, p

The Q-VD-OPH irreversibily binds to activated Caspases to block apoptosis. Apoptosis is a physiological form of cell death, which plays an important role in embryogenesis, cellular homoestasis, tissue atrophy, and removal of damaged and mutated cells.

Caspases, which act as molecular chainsaw, are the central components of apoptosis response. Caspases are Cysteine proteases that cleave after aspartate residue in their substrates. At least 7 of 14 known mammalian caspases have important role in apoptosis.

  • Exceptional alternative to the Fluoromethylketone family of inhibitors (these compounds form the basis for important new drug candidates for a wide variety of disease targets)
  • Nontoxic&mdash:preliminary study showed no evidence of toxicity in mice injected intraperitoneally with more than 1000mg/kg
  • Improved potency—IC50 values against purified caspases are in the nanomolar range; in an in vitro cell based assay, greater than 90% inhibition of apoptosis was achieved)
  • Optimal dilutions should be determined by the laboratory for each specific application
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Quantity 1 mg
Molecular Weight (g/mol) 513.49
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.