missing translation for 'onlineSavingsMsg'
Learn More

Q-VD-OPh, MedChemExpress

Product Code. p-200098798
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16493286

Brand: Medchem Express HY12305

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Q-VD-OPh is an irreversible pan-caspase inhibitor with potent antiapoptotic properties; inhibits caspase 7 with an IC50 of 48 nM and 25-400 nM for other caspases including caspase 1, 3, 8, 9, 10, and 12. Q-VD-OPh can inhibits HIV infection. Q-VD-OPh is able to cross the blood-brain barrier.

Q-VD-OPh is an irreversible pan-caspase inhibitor with potent antiapoptotic properties

Specifications

Chemical Name or Material Q-VD-OPh
CAS 1135695-98-5
Color White
Purity Grade Notes Research
Molecular Formula C26H25F2N3O6
Quantity 25 mg
Synonym QVD-OPH Quinoline-Val-Asp-Difluorophenoxymethylketone
Solubility Information DMSO : 100 mg/mL (194.75 mM; Need ultrasonic)
SMILES O=C(O)C[C@H](NC([C@@H](NC(C1=NC2=CC=CC=C2C=C1)=O)C(C)C)=O)C(COC3=C(F)C=CC=C3F)=O
Molecular Weight (g/mol) 513.49
Formula Weight 513.49
Percent Purity 98.04%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.