missing translation for 'onlineSavingsMsg'
Learn More

Q-VD-OPh, MedChemExpress

Product Code. 16427807
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16427807 5 mg 5mg
16493286 25 mg 25mg
16435088 10 mg 10mg
16401239 1 mg 1mg
16401327 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16427807 Supplier Medchem Express Supplier No. HY12305
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Q-VD-OPh is an irreversible pan-caspase inhibitor with potent antiapoptotic properties; inhibits caspase 7 with an IC50 of 48 nM and 25-400 nM for other caspases including caspase 1, 3, 8, 9, 10, and 12. Q-VD-OPh can inhibits HIV infection. Q-VD-OPh is able to cross the blood-brain barrier.

Q-VD-OPh is an irreversible pan-caspase inhibitor with potent antiapoptotic properties

Specifications

Chemical Name or Material Q-VD-OPh
CAS 1135695-98-5
Color White
Purity Grade Notes Research
Molecular Formula C26H25F2N3O6
Quantity 5 mg
Synonym QVD-OPH Quinoline-Val-Asp-Difluorophenoxymethylketone
Solubility Information DMSO : 100 mg/mL (194.75 mM; Need ultrasonic)
SMILES O=C(O)C[C@H](NC([C@@H](NC(C1=NC2=CC=CC=C2C=C1)=O)C(C)C)=O)C(COC3=C(F)C=CC=C3F)=O
Molecular Weight (g/mol) 513.49
Formula Weight 513.49
Percent Purity 98.04%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.