missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Q-PHOENIX-90 Yellow Safety Storage Cabinets

Product Code. 15890238 Shop All asecos Products
Click to view available options
Configuration:
3 Shelves, 1 Insert, 1 Bottom Collecting Sump
3 Tray Shelves, 1 Perforated Insert (365 mm), 1 Bottom Collecting Sump
4 Drawers
6 Drawers
6 Drawers, 1 Bottom Collecting Sump
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15890238

Brand: asecos™ 3214904033635

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

For the safe and approved storage of flammable hazardous substances in working areas with fire resistance up to 90 minutes.

  • Extremely robust design with scratch-proof surface
  • Space-saving folding doors, ideal for use in case of narrow passage widths in front of the cabinet
  • Continuous anti-trapping door strips on the folding doors offer maximum protection during operation
  • Integrated transport base for simple in-house transport, can be moved upright through doors
  • Adjustable feet installed in the base allow the safe and fast compensation of minor surface unevenness
  • Modern bicolor design and language-free labelling
  • Fire resistant up to 90 minutes
  • Doors lockable with cylinder lock, cabinets can be integrated by the customer in an existing master key system
  • Locking status indicator (red/green) signals the locking status
  • In accordance with EN 14470-1 and TRGS 510
  • Integrated air ducts ready for connection (DN 75) to a technical exhaust system
  • Visual inspection of the exhaust air function in the exhaust grille (rotating fan impeller)
  • Available interior equipment includes drawers, height-adjustable shelves and bottom collecting sumps
  • Triple-hinged doors, safety elements assembled outside the storage compartment for increased protection against corrosion
  • Doors stop in any position and self-close in the event of a fire

Specifications

Certifications/Compliance EN 14470-1, TRGS 510
Configuration 3 Shelves, 1 Insert, 1 Bottom Collecting Sump
Dimensions (D x W x H) Interior 522 x 1050 x 1647 mm
Dimensions (D x W x H) 615 x 1193 x 1953 mm
Door Style Folding
For Use With (Application) Storage of flammable, hazardous substances
Load Bearing Capacity Shelves: 75 kg each, Insert: 75 kg, Sump: 33 L, Maximum load: 600 kg
No. of Doors 2
Weight (Metric) 474.5 kg
Color Anthracite Gray, Warning Yellow
Material Gypsum
Product Type Flammables Storage Cabinet
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.