missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet Q-CLASSIC-90 Winged Doors

Product Code. p-8259003 Shop All asecos Products
Click to view available options
No. of Shelves:
3
6
Color:
Anthracite Gray, Light Gray
Anthracite Gray, Warning Yellow
Weight (Metric):
480.50 kg
537 kg
Unit Size:
Each
This item is not returnable. View return policy

For indoor use only

Product Code. 15950496

Brand: asecos™ 3000104033640

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

For indoor use only

Safe and approved storage of flammable hazardous substances in working areas

  • Safe and approved storage of flammable hazardous substances in working areas
  • Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
  • Easy handling with comfort: doors stop in any position, self-closing in the event of fire
  • No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
  • Easy transport: integrated transport base with removable, optional base cover
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
  • Fire resistance 90 minutes
  • Type tested in accordance with EN 14470-1
  • EN 16121/16122 conformity
  • GS certified
  • CE compliant

Specifications

Certifications/Compliance Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Configuration 3 x tray shelf, 1 x perforated insert (135 mm high), 1 x bottom collecting sump (sheet steel powder-coated)
Depth (Metric) Exterior 615 mm
Depth (Metric) Interior 522 mm
Description Warning Yellow doors
Dimensions (D x W x H) Interior 522 x 1050 x 1647 mm
Dimensions (D x W x H) 615 x 1193 x 1953 mm
Door Style Wing
For Use With (Application) Safe and approved storage of flammable hazardous substances in working areas
Height (Metric) Exterior 1953 mm
Height (Metric) Interior 1647 mm
Model Q90.195.120
No. of Doors 2
No. of Shelves 3
Shelf Type Tray shelf
Surface Coating Powder-coated textured
Weight (Metric) 480.50 kg
Width (Metric) Exterior 1193 mm
Width (Metric) Interior 1050 mm
Color Anthracite Gray, Warning Yellow
Height (Metric) Opening 90 mm
Material Steel
Product Type Flammables Storage Cabinet
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.