missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 30 Safety Storage Cabinet Q-CLASSIC-30 with 2 Doors

Product Code. 15986486 Shop All asecos Products
Click to view available options
Configuration:
3 x Shelf, 1 x Perforated Insert (375 mm), 1 x Bottom Collecting Sump
3 x Shelf, 1 x Perforated Insert (380 mm), 1 x Bottom Collecting Sump
3 x Shelf, 1 x Perforated Insert, 1 x Bottom Collecting Sump
4 x Drawer
6 x Drawer
Color:
Anthracite Gray
Anthracite Gray, Light Gray Door
Anthracite Gray, Warning Yellow
Anthracite Gray, Warning Yellow Door
Weight (Metric):
271 kg
271.50 kg
302 kg
334 kg
Unit Size:
Each
This item is not returnable. View return policy

For indoor use only

Product Code. 15986486

Brand: asecos™ 3100306133635

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

For indoor use only

Safe and approved storage of flammable hazardous substances in working areas

  • Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
  • Easy handling with comfort: doors stop in any position, self-closing in the event of fire
  • No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
  • Easy transport: integrated transport base with removable, optional base cover or adjusting aids with base cover
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system

Warranty

10 years

Specifications

Certifications/Compliance Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Configuration 3 x Shelf, 1 x Perforated Insert, 1 x Bottom Collecting Sump
Depth (Metric) Exterior 620 mm
Depth (Metric) Interior 519 mm
Dimensions (D x W x H) Interior 519 x 1050 x 1626 mm
Dimensions (D x W x H) 620 x 1164 x 1947 mm
Door Style Wing
For Use With (Application) Safe and approved storage of flammable hazardous substances in working areas
Height (Metric) Exterior 1947 mm
Height (Metric) Interior 1626 mm
Model Q30.195.116
No. of Doors 2
No. of Shelves 3
Shelf Type Standard
Surface Coating Powder-coated textured
Weight (Metric) 271.50 kg
Width (Metric) Exterior 1164 mm
Width (Metric) Interior 1050 mm
Color Anthracite Gray, Light Gray Door
Height (Metric) Opening 90 mm
Material Steel
Product Type Flammables Storage Cabinet
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.