missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ Q-CLASSIC-30 One-Door Yellow Safety Storage Cabinets

Product Code. 15863354 Shop All asecos Products
Change view
Click to view available options
Load Bearing Capacity:
Shelves 75 kg, Insert 75 kg, Sump 22 L, Max load 600 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Load Bearing Capacity Unit Size
15863354 Shelves 75 kg, Insert 75 kg, Sump 22 L, Max load 600 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15863354 Supplier Asecos™ Supplier No. 3100806033622
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the safe and approved storage of flammable hazardous substances in working areas with fire resistance up to 30 minutes.

  • The sophisticated design and use of melamine-resin-coated, fireproof special plates ensures an optimum price/performance ratio
  • Integrated transport base for simple in-house transport, can be transported upright through doors
  • Adjustable feet installed in the base allow the safe and fast compensation of minor surface unevenness
  • Modern bicolor design and language-free labelling for optimum international usability
  • Door lockable with cylinder lock, and can be integrated by the customer in an existing locking system
  • Interior equipment such as drawers, height-adjustable shelves and bottom collecting sumps available
  • Fire resistant up to 30 minutes
  • Triple-hinged doors, stoppable in any position, self-closing in the event of fire
  • Integrated air ducts ready for connection (DN 75) to a technical exhaust system for ventilation
  • In accordance with EN 14470-1 and TRGS 510

Specifications

Certifications/Compliance EN 14470-1, TRGS 510
Configuration 3 x 75 kg powder-coated sheet steel shelves; 1 x 75 kg powder-coated sheet steel insert; 1 x 22 L powder-coated sheet steel bottom collecting sump
Dimensions (D x W x H) Interior 519 x 750 x 1626 mm
Dimensions (D x W x H) 620 x 864 x 1947 mm
Door Style Wing
For Use With (Application) Storage of flammable and hazardous materials
Load Bearing Capacity Shelves 75 kg, Insert 75 kg, Sump 22 L, Max load 600 kg
No. of Doors 1
Surface Coating Plastic laminated
Weight (Metric) 224 kg
Color Yellow
Material Melamine Resin
Product Type Flammables Storage Cabinet
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.