missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PYY Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5110763

Product Code. 17188683

  • £550.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PIKPEAPGED ASPEELNRYY ASLRHYLNLV TRQRYGKRDG PDRLLSKTFF PDGEDRPVRS RSEGPDLW Highest antigen sequence indentity to the following orthologs: Mouse - 73%, Rat - 74%.

Peptide tyrosine-tyrosine amide (3-36), or PYY (3-36), is a major metabolite of the gut hormone PYY and is produced by the action of dipeptidyl peptidase IV on PYY in the intestinal brush border and in the circulation. PYY (3-36) is a potent inhibitor of food intake in rats and humans, acting selectively on the Y2 receptor to inhibit orexigenic NPY neurons in the arcuate nucleus, thus disinhibiting anorexigenic POMC neurons. The anorexigenic effect of PYY (3-36) is additive to that of the co-secreted gut hormone GLP-1.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PYY
Polyclonal
Unconjugated
Pyy
GHYY; Peptide tyrosine tyrosine; peptide tyrosine-tyrosine (YY); peptide YY; peptide YY (mapped); Peptide YY(3-36); Peptide YY3-36; Peptide YY3-37; peptide-YY; prepro-PYY; PYY; PYY II; PYY1; PYY-1; PYY-2; PYYI; PYY-I; PYYII; PYY-II; RATGHYY; Yy
Rabbit
Antigen affinity chromatography
RUO
5697
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P10082
Pyy
Recombinant Protein corresonding to Human PYY. Recombinant protein control fragment (Product #RP-90212).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.