missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic aldehyde dimethyl acetal, 98%

Product Code. 10654852 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10654852

Brand: Thermo Scientific Acros 173805000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6342-56-9 | C5H10O3 | 118.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6342-56-9
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00008758
InChI Key ULVSHNOGEVXRDR-UHFFFAOYSA-N
Synonym 1,1-dimethoxyacetone, pyruvic aldehyde dimethyl acetal, 2-propanone, 1,1-dimethoxy, methylglyoxal dimethyl acetal, 1,1-dimethoxy-2-propanone, dimethoxymethyl methyl ketone, pyruvaldehyde dimethyl acetal, 1,1-dimethoxy acetone, dimethoxyacetone, pyruvaldehyde, 1-dimethyl acetal
PubChem CID 80650
IUPAC Name 1,1-dimethoxypropan-2-one
SMILES CC(=O)C(OC)OC

Specifications

Melting Point -57.0°C
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 143.0°C to 147.0°C
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCH(OCH3)2
Refractive Index 1.3970 to 1.3990
Quantity 500 g
Beilstein 01, IV, 3631
Specific Gravity 1
Solubility Information Solubility in water: 350g/L (20°C). Other solubilities: miscible with methanol, acetone and thf
Viscosity 0.85 mPa.s (25°C)
Formula Weight 118.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Pyruvic aldehyde dimethyl acetal, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pyruvic aldehyde dimethyl acetal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.